Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRMT2A Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505555S


Description
TRMT2A Monoclonal antibody specifically detects TRMT2A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
TRMT2A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BX59 | |
Mouse | |
Affinity chromatography | |
RUO | |
55080 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C3 | |
Unconjugated | |
TAPBPL | |
HpaII tiny fragments locus 9c; hpaII tiny fragments locus 9c protein; Htf9c; Htf9-c; TRM2 tRNA methyltransferase 2 homolog A; TRM2 tRNA methyltransferase 2A; TRMT2A; tRNA (uracil-5-)-methyltransferase homolog A; tRNA methyltransferase 2 homolog A; tRNA methyltransferase 2 homolog A (S. cerevisiae) | |
Full length human recombinant protein of human TAPBPL produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction