Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT2A Recombinant Rabbit Monoclonal Antibody (JE55-60)
GREENER_CHOICE

Catalog No. PIMA542495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542495 Supplier Invitrogen™ Supplier No. MA542495
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: 293 cell lysate, Jurkat cell lysate, MCF-7 cell lysate, human liver carcinoma tissue, human thyroid tissue, human breast carcinoma tissue. Subcellular Location: Nucleoplasm and Cytosol.(Predicted).

The protein encoded by TRMT2A is of unknown function. However, it is orthologous to the mouse Trmt2a gene and contains an RNA methyltransferase domain. Expression of TRMT2A varies during the cell cycle, with aberrant expression being a possible biomarker in certain breast cancers. Several transcript variants encoding two different isoforms have been found for TRMT2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE55-60
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TRMT2A
Gene Accession No. Q8IZ69
Gene Alias HpaII tiny fragments locus 9c; hpaII tiny fragments locus 9c protein; Htf9c; Htf9-c; TRM2 tRNA methyltransferase 2 homolog A; TRM2 tRNA methyltransferase 2A; TRMT2A; tRNA (uracil-5-)-methyltransferase homolog A; tRNA methyltransferase 2 homolog A; tRNA methyltransferase 2 homolog A (S. cerevisiae)
Gene Symbols TRMT2A
Host Species Rabbit
Immunogen Recombinant fragment within N-terminal Human HTF9C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.