Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT2B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen TRMT2B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155171
SDP
View Documents
Novus Biologicals
NBP155171
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

TRMT2B Polyclonal specifically detects TRMT2B in Human samples. It is validated for Western Blot.
Specifications

Specifications

TRMT2B
Polyclonal
Purified
RUO
chromosome X open reading frame 34, CXorf34, dJ341D10.3, EC 2.1.1.35, FLJ12687, TRM2 homolog, TRM2 tRNA methyltransferase 2 homolog B (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog
TRMT2B
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
Q96GJ1
79979
Synthetic peptides corresponding to CXORF34 The peptide sequence was selected from the middle region of CXORF34. Peptide sequence GAACGLTSLYFQESTMTRCSHQQSPYQLLFGEPYIFEELLSLKIRISPDA.
Primary
56 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.