Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551997 Supplier Invitrogen™ Supplier No. PA551997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSLAQHTSRV LGIELLEQAV EDARWTAAFN GITNSEFHTG QAEKILPGLL KSKEDGQSIV AVVNPARAGL HYKVIQAIRN FRAIHTLVFV SCKLHGESTR NVIELCCPPD PAKKLLGEPF VLQQAVPVDL FPHT Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 47%.

Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRMT2B
Gene Accession No. Q96GJ1
Gene Alias CXorf34; dJ341D10.3; TRM2 homolog; TRM2 tRNA methyltransferase 2 homolog B; TRMT2B; tRNA (uracil(54)-C(5))-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog; tRNA methyltransferase 2 homolog B
Gene Symbols TRMT2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRMT2B. Recombinant protein control fragment (Product #RP-102535).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.