Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120866 Supplier Invitrogen™ Supplier No. PA5120866
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Raji, U-87MG, SW480, MCF7, NCI-H460. Immunogen sequence: MAGLKRRVPL HSLRYFISMV GLFSKPGLLP WYARNPPGWS QLFLGTVCKG DFTRVIATKC QKGQKSQKKP SHLGPLDGSW QERLADVVTP LWRLSYEEQL KVKFAAQKKI LQRLESYIQM LNGVSVTTAV PKSERLSCLL HPIIPSPVIN GYRNKSTFSV NRGPDGNPKT VGFYLGTWRD GNVVCVQSNH LKNIPEKHSQ VAQYYEVFLR QSPLEPCLVF HEGGYWRELT VRTNSQGHTM AIITFHPQKL.

Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT2B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRMT2B
Gene Accession No. Q96GJ1
Gene Alias CXorf34; dJ341D10.3; TRM2 homolog; TRM2 tRNA methyltransferase 2 homolog B; TRMT2B; tRNA (uracil(54)-C(5))-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog; tRNA methyltransferase 2 homolog B
Gene Symbols TRMT2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TRMT2B (NP_079193.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 503462, 79979
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.