Learn More
Invitrogen™ TRMT5 Monoclonal Antibody (OTI1B5)

Description
TRMT5 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TRMT5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TRMT5 |
| Gene Accession No. | Q32P41 |
| Gene Alias | 2610027O18Rik; COXPD26; hypothetical protein LOC528221; KIAA1393; M1G-methyltransferase; mKIAA1393; RGD1306567; TRM5; TRM5 tRNA methyltransferase 5; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine(37)-N1)-methyltransferase; LOW QUALITY PROTEIN: tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase-like protein; tRNA [GM37] methyltransferase; tRNA methyltransferase 5; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.