Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569558 Supplier Thermo Scientific Supplier No. PA569558
Add to Cart
Add to Cart

TRMT5 Polyclonal Antibody

TRMT5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRMT5
Gene Accession No. Q9D0C4
Gene Alias 2610027O18Rik; COXPD26; hypothetical protein LOC528221; KIAA1393; M1G-methyltransferase; mKIAA1393; RGD1306567; TRM5; TRM5 tRNA methyltransferase 5; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine(37)-N1)-methyltransferase; LOW QUALITY PROTEIN: tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase-like protein; tRNA [GM37] methyltransferase; tRNA methyltransferase 5; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Symbols TRMT5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TRMT5 (aa 256-305).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76357
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.