Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRMT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561409
Description
Immunogen sequence: KMIVMETCAG LVLGAMMERM GGFGSIIQLY PGGGPVRAAT ACFGFPKSFL SGLYEFPLNK VDSLLHGTFS AKMLSSEPKD SALVEESNGT LE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
Specifications
TRMT6 | |
Polyclonal | |
Unconjugated | |
TRMT6 | |
3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; Kiaa1153; MGC5029; mKIAA1153; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; RGD1308877; Trm6; TRMT6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.40 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJA5 | |
TRMT6 | |
Recombinant protein corresponding to Human TRMT6. Recombinant protein control fragment (Product #RP-99317). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction