Learn More
Invitrogen™ TRMT6 Polyclonal Antibody

Description
Immunogen sequence: LKDKGIKGEEI VQQLIENSTT FRDKTEFAQD KYIKKKKKKY EAIITVVKPS TRILSIMYYA REPGKINHMR YDTLAQMLTL GNIRAGN Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Specifications
Specifications
| Antigen | TRMT6 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRMT6 |
| Gene Accession No. | Q9UJA5 |
| Gene Alias | 3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; Kiaa1153; MGC5029; mKIAA1153; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; RGD1308877; Trm6; TRMT6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6 |
| Gene Symbols | TRMT6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.