Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597194 Supplier Invitrogen™ Supplier No. PA597194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEGSGEQPGP QPQHPGDHRI RDGDFVVLKR EDVFKAVQVQ RRKKVTFEKQ WFYLDNVIGH SYGTAFEVTS GGSLQPKKKR EEPTAETKEA GTDNRNIVDD GKSQKLTQDD IKALKDKGIK GEEIVQQLIE NSTTFRDKTE FAQDKYIKKK KKKYEAIITV VKPSTRILSI MYYAREPGKI NHMRYDTLAQ MLTLGNIRAG; Positive Samples: U-87MG, HepG2; Cellular Location: Nucleus.

Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRMT6
Gene Accession No. Q9UJA5
Gene Alias 3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; Kiaa1153; MGC5029; mKIAA1153; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; RGD1308877; Trm6; TRMT6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6
Gene Symbols TRMT6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TRMT6 (NP_0570232).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51605
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.