Learn More
Invitrogen™ TRMT61B Polyclonal Antibody

Description
Immunogen sequence: KRGTAITFPK DINMILSMMD INPGDTVLEA GSGSGGMSLF LSKAVGSQGR VISFEVRKDH HDLAKKNYKH WRDSWKLSHV EEWPDNVDFI HKDISGATED IKSLTFDAVA LDMLN Highest antigen sequence identity to the following orthologs: Mouse - 22%, Rat - 24%.
Specifications
Specifications
| Antigen | TRMT61B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRMT61B |
| Gene Accession No. | Q9BVS5 |
| Gene Alias | 1110039F03Rik; 623451; Gm6431; mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B; tRNA methyltransferase 61B |
| Gene Symbols | TRMT61B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.