Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRMU Mouse anti-Canine, Human, Mouse, Clone: OTI2A4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA505699

Description
Catalyzes the 2-thiolation of uridine at the wobble position (U34) of mitochondrial tRNA(Lys), tRNA(Glu) and tRNA(Gln). Required for the formation of 5-taurinomethyl-2- thiouridine (tm5s2U) of mitochondrial tRNA(Lys), tRNA(Glu), and tRNA(Gln) at the wobble position. ATP is required to activate the C2 atom of the wobble base.Specifications
TRMU | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75648, Q9DAT5 | |
TRMU | |
Full length human recombit protein of human TRMU produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A4 | |
Unconjugated | |
TRMU | |
LCAL3, MTO2, MTU1, TRMT, TRMT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
481204, 55687, 72026 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction