Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRMU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110618
Description
Immunogen sequence: EDNKVLGTHK GWFLYTLGQR ANIGGLREPW YVVEKDSVKG DVFVAPRTDH PALYRDLLRT SRVHWIAEEP PAALVRDKMM ECHFRFRHQM ALVPCVLTLN QDGTVWVTAV QAVRALATGQ FAVFYKGDEC LGS.
Catalyzes the 2-thiolation of uridine at the wobble position (U34) of mitochondrial tRNA(Lys), tRNA(Glu) and tRNA(Gln). Required for the formation of 5-taurinomethyl-2- thiouridine (tm5s2U) of mitochondrial tRNA(Lys), tRNA(Glu), and tRNA(Gln) at the wobble position. ATP is required to activate the C2 atom of the wobble base.
Specifications
TRMU | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75648 | |
TRMU | |
Recombinant Protein corresonding to Human TRMU. Recombinant protein control fragment (Product #RP-108303). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
TRMU | |
1110005N20Rik; 1600025P05Rik; AI314320; LCAL3; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; mitochondrial tRNA-specific 2-thiouridylase 1; MTO2; MTO2 homolog; MTU1; TRMT; Trmt1; Trmu; tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase 1; tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase; tRNA mitochondrial 2-thiouridylase; TRNT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55687 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction