Learn More
Invitrogen™ TRMU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113877
Description
Immunogen sequence: ALATGQFAVF YKGDECLGSG KILRLGPSAY TLQKGQRRAG MATESPSDSP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Catalyzes the 2-thiolation of uridine at the wobble position (U34) of mitochondrial tRNA(Lys), tRNA(Glu) and tRNA(Gln). Required for the formation of 5-taurinomethyl-2- thiouridine (tm5s2U) of mitochondrial tRNA(Lys), tRNA(Glu), and tRNA(Gln) at the wobble position. ATP is required to activate the C2 atom of the wobble base.
Specifications
TRMU | |
Polyclonal | |
Unconjugated | |
TRMU | |
1110005N20Rik; 1600025P05Rik; AI314320; LCAL3; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; mitochondrial tRNA-specific 2-thiouridylase 1; MTO2; MTO2 homolog; MTU1; TRMT; Trmt1; Trmu; tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase 1; tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase; tRNA mitochondrial 2-thiouridylase; TRNT1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55687 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75648 | |
TRMU | |
Synthetic peptide directed towards the C terminal region of human TRMU (aa 362-411). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.