Learn More
Thermo Scientific™ tRNA for LightShift™ Chemiluminescent RNA EMSA Kit PROMO
Description
The RNA EMSA Kit uses biotinylated RNA probes and a chemiluminescent substrate system to achieve fast and safe detection of RNA-protein complexes with sensitivity equivalent to traditional 32P-isotopic methods. The complete kit comes with all reagents required to set up and optimize protein-RNA binding conditions, a positive control for protein-RNA interactions and reagents for chemiluminescent detection of the nucleic acid interaction.
Highlights:
Sensitive – chemiluminescent detection comparable to radioactive detection
Time-saving – develop X-ray films after 1- to 5-minute exposures, versus 16 hour exposures needed with radioactive systems
Flexible – compatible with RNA probes biotinylated by different methods
Easy to use – complete kit includes optimized reagents for binding reactions and RNA probe detection
Non-radioactive – eliminate waste concerns from radioactive RNA probes
Specifications
Specifications
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -70°C. |
Detection Method | Chemiluminescence |
Format | Kit |
Product Line | LightShift™ |
Product Type | tRNA for LightShift RNA EMSA Kit |
Quantity | 100 μL |
Target Specificity | Not Target-Specific |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.