Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRNP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144259
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144259 Supplier Invitrogen™ Supplier No. PA5144259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRNP1 Polyclonal Antibody for IHC (P), ELISA

DNA-binding factor that regulates the expression of a subset of genes and plays a key role in tangential, radial, and lateral expansion of the brain neocortex. Regulates neural stem cells proliferation and the production of intermediate neural progenitors and basal radial glial cells affecting the process of cerebral cortex gyrification. May control the proliferation rate of cells by regulating their progression through key cell-cycle transition points.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRNP1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.215 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TRNP1
Gene Accession No. Q6NT89
Gene Alias C1orf225; TMF1-regulated nuclear protein 1; TMF-regulated nuclear protein 1; TNRP; TRNP; TRNP1
Gene Symbols TRNP1
Host Species Rabbit
Immunogen Recombinant Human TMF-regulated nuclear protein 1 protein (10-180AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388610
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.