Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110524 Supplier Invitrogen™ Supplier No. PA5110524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAEVEFTTDW QKDAERRDLT INSMFLGFDG TLFDYFNGYE DLKNKKVRFV GHAKQRIQED YLRILRYFRF YGRIVDKPGD HDPETLEAIA ENAKGLAGIS GERIWVELKK ILVGNHVNHL IHLIYDLDVA PYIGLPANAS LEEFDKVSKN VDGFSPKPVT LLASLFKVQD DVTKLDLRLK IAKEEKNLGL FIVKNRKDLI KATDSSDPLK PYQDFIIDSR EPDATTRVCE LLKYQGEHCL LKEMQQWSIP PFPVSGHDIR KVGISSGKEI GALLQQLREQ WKKSGYQMEK DELLSYIKKT.

TRNT1 belongs to the tRNA nucleotidyltransferase/poly(A) polymerase family. It adds and repairs the conserved 3'-CCA sequence necessary for the attachment of amino acids to the 3' terminus of tRNA molecules, using CTP and ATP as substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRNT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRNT1
Gene Accession No. Q8K1J6, Q96Q11
Gene Alias 2410043H24Rik; 2610044E04Rik; 9830143O18Rik; AA408384; ATP(CTP):tRNA nucleotidyltransferase; bMtCCA; C76540; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA1; CCA-adding enzyme; CGI-47; mitochondrial CCA-adding tRNA-nucleotidyltransferase; mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; MtCCA; mt-Trat; RPEM; SIFD; tRNA adenylyltransferase, mitochondrial; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA nucleotidyl transferase-like; tRNA-nucleotidyltransferase; tRNA-nucleotidyltransferase 1 mitochondrial-like protein; tRNA-nucleotidyltransferase 1, mitochondrial; TRNT1; Unknown (protein for MGC:140399); zgc:86645
Gene Symbols TRNT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 135-434 of human TRNT1 (NP_886552.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312616, 51095, 70047
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.