Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRNT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541859
Description
Peptide sequence: PQDIDFATTA TPTQMKEMFQ SAGIRMINNR GEKHGTITAR LHEENFEITT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 93%.
TRNT1 belongs to the tRNA nucleotidyltransferase/poly(A) polymerase family. It adds and repairs the conserved 3'-CCA sequence necessary for the attachment of amino acids to the 3' terminus of tRNA molecules, using CTP and ATP as substrates.
Specifications
TRNT1 | |
Polyclonal | |
Unconjugated | |
TRNT1 | |
2410043H24Rik; 2610044E04Rik; 9830143O18Rik; AA408384; ATP(CTP):tRNA nucleotidyltransferase; bMtCCA; C76540; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA1; CCA-adding enzyme; CGI-47; mitochondrial CCA-adding tRNA-nucleotidyltransferase; mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; MtCCA; mt-Trat; RPEM; SIFD; tRNA adenylyltransferase, mitochondrial; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA nucleotidyl transferase-like; tRNA-nucleotidyltransferase; tRNA-nucleotidyltransferase 1 mitochondrial-like protein; tRNA-nucleotidyltransferase 1, mitochondrial; TRNT1; Unknown (protein for MGC:140399); zgc:86645 | |
Rabbit | |
Protein A | |
RUO | |
51095 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96Q11 | |
TRNT1 | |
Synthetic peptide directed towards the N-terminal of human TRNT1 (aa 75-124). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction