Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRO Polyclonal Antibody

Catalog No. PIPA572872
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572872 Supplier Invitrogen™ Supplier No. PA572872

Rabbit Polyclonal Antibody

TRO Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a membrane protein that mediates cell adhesion between trophoblastic cells and the epithelial cells of the endometrium. The encoded protein participates in cell signalling during embryo implantation, and may also be involved in cancer formation. This gene is located near several other closely related genes on chromosome X. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRO
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRO
Gene Accession No. Q12816
Gene Alias AA409408; KIAA1114; LOC100912141; MAGE superfamily protein; MAGED3; MAGE-d3; MAGE-D3 antigen; Maged3l; magphinin; melanoma antigen, family D, 3; necdin and trophinin like; Tnn; TRO; Trol; trophinin; trophinin-2; trophinin-like
Gene Symbols TRO
Host Species Rabbit
Immunogen Trophinin antibody for a 16 amino acid peptide near the amino terminus of human Trophinin. The immunogen is within amino acids 390 - 440 of Trophinin.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912141, 56191, 7216
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.