Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRO Polyclonal Antibody

TRO Polyclonal Antibody
Supplier: Thermo Scientific PA545460
Description
TRO Polyclonal Antibody for Western Blot
Specifications
| TRO | |
| Polyclonal | |
| Unconjugated | |
| TRO | |
| AA409408; KIAA1114; LOC100912141; MAGE superfamily protein; MAGED3; MAGE-d3; MAGE-D3 antigen; Maged3l; magphinin; melanoma antigen, family D, 3; necdin and trophinin like; Tnn; TRO; Trol; trophinin; trophinin-2; trophinin-like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7216 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q12816 | |
| TRO | |
| Synthetic peptide directed towards the C-terminal of human TRO. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction