Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trolamine, NF, 99-107.4%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Description
Spectrum™ Chemical Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Ignition Residue | 0.0005 |
Packaging | Amber Glass Bottle |
Refractive Index | 1.481-1.486 |
Quantity | 4 L |
Percent Purity | 99 to 107.4% |
Grade | NF |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction