Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tromethamine, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18607615
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

TR149, 77-86-1, C4H11NO3

Spectrum™ Chemical Tromethamine, USP is used to treat metabolic acidosis. All Spectrum Chemical USP products, including all USP tromethamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

CAS Max % 1
pH 10.0 to 11.5
Ignition Residue 0.001
Loss on Drying 0.01
Melting Range 168-172°C
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 99 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.