Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROP-2 Antibody (020), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289493R
Description
TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TROP-2 | |
Monoclonal | |
DyLight 550 | |
Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TROP-2 (NP_002344.2; Met1-Thr274). | |
0.1 mL | |
Signal Transduction | |
4070 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
20 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction