Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trop-2 Mouse, anti-Human, R718, Clone: 162-46, BD OptiBuild™

Catalog No. BDB752129 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752129 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752129 Supplier BD Biosciences Supplier No. 752129
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 162-46 monoclonal antibody recognizes human Trop-2 which is also known as GA733-1. Trop-2 is encoded by TACSTD2 (tumor-associated calcium signal transducer 2). It is a single-pass type-1 transmembrane glycoprotein. Trop-2 was originally identified to be expressed on human trophoblast and choriocarcinoma cell lines. Trop-2 is expressed in most human carcinomas. Trop-2 appears to be involved in the transduction of an intracellular calcium signal, although the exact physiological mechanism has yet to be elucidated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Trop-2
Applications Flow Cytometry
Classification Monoclonal
Clone 162-46
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.