Learn More
Invitrogen™ TROP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121175
Description
Positive test controls include: BxPC-3, A-431, Mouse kidney. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: HTAAQDNCTC PTNKMTVCSP DGPGGRCQCR ALGSGMAVDC STLTSKCLLL KARMSAPKNA RTLVRPSEHA LVDNDGLYDP DCDPEGRFKA RQCNQTSVCW CVNSVGVRRT DKGDLSLRCD ELVRTHHILI DLRHRPTAGA FNHSDLDAEL RRLFRERYRL HPKFVAAVHY EQPTIQIELR QNTSQKAAGD VDIGDAAYYF ERDIKGESLF QGRGGLDLRV RGEPLQVERT LIYYLDEIPP KFSM.
TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.
Specifications
| TROP2 | |
| Polyclonal | |
| Unconjugated | |
| TACSTD2 | |
| 40kD glycoprotein, identified by monoclonal antibody GA733; C80403; cell surface glycoprotein TROP2; cell surface glycoprotein Trop-2; EGP1; EGP-1; epithelial glycoprotein-1; GA733; GA7331; GA733-1; gastrointestinal tumor-associated antigen GA7331; GP50; Ly97; lymphocyte antigen 97; M1S1; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; pancreatic carcinoma marker protein GA7331; pancreatic carcinoma marker protein GA733-1; parturition-related protein 1; Prp1; TACD2; TACSTD2; Trop2; truncated TACSTD2; tumor-associated calcium signal transducer 2; tunor-associated calcium signal transducer 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4070, 494343, 56753 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09758, Q6P9Z6, Q8BGV3 | |
| TACSTD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-270 of human TROP-2 (NP_002344.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.