Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropine, 98%

Catalog No. p-3737709
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 120-29-6 | C8H15NO | 141.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-29-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00005551
InChI Key CYHOMWAPJJPNMW-RNFRBKRXSA-N
Synonym tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
PubChem CID 6101956
IUPAC Name (1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1C2CCC1CC(C2)O

Specifications

Melting Point 62.0°C to 64.0°C
Color White to Yellow
Density 1.0400g/mL
Boiling Point 233.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 16
Specific Gravity 1.04
Merck Index 15, 9960
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in ether and chloroform, freely soluble in alcohol
Formula Weight 141.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tropine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/
EINECSNumber 204-384-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.