Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropinone, 99%

Catalog No. AAL0201114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0201114 25 g
AAL0201106 5 g
AAL0201122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0201114 Supplier Thermo Scientific Chemicals Supplier No. L0201114
Add to Cart

CAS: 532-24-1 | C8H13NO | 139.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-24-1
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.20
MDL Number MFCD00005549
InChI Key QQXLDOJGLXJCSE-UHFFFAOYNA-N
Synonym tropinone, 8-methyl-8-azabicyclo 3.2.1 octan-3-one, 3-tropanone, tropanone, tropanon, tropinon, tropionone, 3-tropinone, 8-azabicyclo 3.2.1 octan-3-one, 8-methyl, 1.alpha.h,5.alpha.h-tropan-3-one
PubChem CID 79038
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-one
SMILES CN1C2CCC1CC(=O)C2

Specifications

Melting Point 40°C to 43°C
Boiling Point 227°C
Flash Point 90°C (194°F)
Odor Odorless
Quantity 25 g
Beilstein 2329
Formula Weight 139.2
Percent Purity 99%
Chemical Name or Material Tropinone
Hazard Category H302-H314
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 208-530-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.