Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tropisetron hydrochloride
GREENER_CHOICESDP

Catalog No. 245950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-595-0 50 mg
24-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-595-0 Supplier R&D Systems™ Supplier No. 2459/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent 5-HT3 antagonist; also α7 nAChR partial agonist

Potent, orally active 5-HT3 receptor antagonist. Antiemetic. Also partial agonist of α7 nAChR. Activates α7Q79G-GlyR chimeric ion channels (EC50 = 38 nM)

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 320.82
Purity >99%
Molecular Weight (g/mol) 320.817
Chemical Name or Material Tropisetron hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.