Learn More
Invitrogen™ Tropomodulin 2 Recombinant Rabbit Monoclonal Antibody (JE53-06)

Description
Tropomodulin 2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | Tropomodulin 2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JE53-06 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Tmod2 |
| Gene Accession No. | P70566, Q9JKK7, Q9NZR1 |
| Gene Alias | neural tropomodulin; neuronal tropomodulin; NTMOD; N-TMOD; TMOD2; tropomodulin 2; tropomodulin 2 (neuronal); tropomodulin-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.