Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin-1, Rat anti-Mouse, Human, Clone: 7F8B8, Millipore Sigma™

Catalog No. MABT1365100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1365100 100 μL
MABT136525 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT1365100 Supplier MilliporeSigma Supplier No. MABT1365100UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Tropomyosin-1, clone 7F8B8, Cat. No. MABT1365, is a rat monoclonal antibody that detects Tropomyosin-1 and has been tested for use in Immunocytochemistry and Western Blotting.

Tropomyosin alpha-1 chain (UniProt: P09493; also known as Alpha-tropomyosin, Tropomyosin-1) is encoded by the TPM1 (also known as C15orf13, TMSA) gene (Gene ID: 7168) in human. Tropomyosin-1 is a cytoskeletal protein that associates with F-actin stress fibers. It binds to actin filaments in muscle and non-muscle cells and plays a central role, in association with the troponin complex, in the calcium dependent regulation of vertebrate striated muscle contraction. It stabilizes actin filaments and regulates the interaction of actin filament with other actin-binding proteins. In non-muscle cells is implicated in stabilizing cytoskeleton actin filaments. Tropomyosin-1 is a homodimeric protein, but can form heterodimer of an alpha (TPM1, TPM3, or TPM4) and a beta (TPM2) chain. Mutations in TPM1 gene have been linked to cardiomyopathy characterized by ventricular hypertrophy and ventricular dilation and impaired systolic function that results in congestive heart failure and arrhythmia. Ten different isoforms of Tropomyosin-1 have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin-1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7F8B8
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected Tropomyosin-1 in Immunocytochemistry applications (Brayford, S., et. al. (2016). Curr Biol. 26(10):1312-8).
Formulation Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Tropomyosin alpha-1 chain;Alpha-tropomyosin;Tropomyosin-1
Gene Symbols TPM1;C15orf13;TMSA
Host Species Rat
Immunogen KLH-conjugated linear peptides corresponding to 24 amino acids from the N-terminal region of isoform 5 and 4 amino acids from the N-terminal region of isoform 2 of human tropomyosin-1.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_000357.3
Test Specificity Clone 7F8B8 detects human and murine Tropomyosin-1. It targets an epitope with in the N-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.