Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tropomyosin-2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299519PE
Description
tropomyosin-2 Polyclonal antibody specifically detects tropomyosin-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
tropomyosin-2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
distal, type 1, tropomyosin 2 (beta) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human tropomyosin-2. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
7169 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction