Learn More
Description
Specifications
Specifications
| Antigen | Tropomyosin-4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2D2A12 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of rat Tropomysin-4 corresponding to exon 1b in Tpm4 gene. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.