Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin alpha-1 Mouse anti-Rat, Clone: 2F4E11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT133725UG
Description
Tropomyosin alpha-1 Monoclonal antibody specifically detects Tropomyosin alpha-1 in Rat samples. It is validated for Western Blotting.
Specifications
Tropomyosin alpha-1 | |
Monoclonal | |
1 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide | |
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Tropomyosin alpha-1 chain, isoform 4. | |
25 μg | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Western Blot | |
2F4E11 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction