Learn More
Tropomyosin alpha-1 Mouse anti-Rat, Mouse, Clone: 3F5E4, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tropomyosin alpha-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3F5E4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of rat Tropomyosin alpha-1 chain, isoform 2. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.