Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin alpha-1 Mouse anti-Rat, Mouse, Clone: 3F5E4, MilliporeSigma™

Catalog No. MABT133825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT133825 25 μL
MABT1338100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT133825 Supplier MilliporeSigma Supplier No. MABT133825UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tropomyosin alpha-1 Monoclonal antibody specifically detects Tropomyosin alpha-1 in Rat, Mouse samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin alpha-1
Applications Western Blot
Classification Monoclonal
Clone 3F5E4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of rat Tropomyosin alpha-1 chain, isoform 2.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.