Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin C (cardiac) Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299435UV
Description
Troponin C (cardiac) Polyclonal specifically detects Troponin C (cardiac) in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Troponin C (cardiac) | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse Troponin C (cardiac). | |
0.1 mL | |
Cell Biology, Cellular Markers | |
7134 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction