Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin C (cardiac) Antibody (SR1283), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32198550UL
Description
Troponin C (cardiac) Monoclonal antibody specifically detects Troponin C (cardiac) in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Troponin C (cardiac) | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1283 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
A synthesized peptide derived from human Troponin C (cardiac) (Uniprot #: P63316) | |
50 μg | |
Cell Biology, Cellular Markers | |
7134 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction