Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | Troponin I |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121541
|
Invitrogen™
PA5121541 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5121540
|
Invitrogen™
PA5121540 |
100 μL |
Each for $511.50
|
|
|||||
Description
Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. The TnI-fast and TnI-slow genes are expressed in fast-twitch and slow-twitch skeletal muscle fibers, respectively, while the TnI-cardiac gene is expressed exclusively in cardiac muscle tissue. This gene encodes the Troponin-I-skeletal-slow-twitch protein. This gene is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The encoded protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.Specifications
Troponin I | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P19237, P19429, P48787, Q9WUZ5 | |
21952, 21954, 7135, 7137 | |
Met1-Asp103 linked with Leu142-Gln187 (Accession P19237), with N-terminal His Tag. | |
Primary | |
-20°C |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
2700018B22Rik; AI747285; Cardiac troponin I; cardiomyopathy, dilated 2A (autosomal recessive); CMD1FF; CMD2A; CMH7; cTNI; DKFZp451O223; RCM1; slow skeletal troponin I isoform; ssTnI; Tn1; TnI; TNN1; TNNC1; TNNI1; TNNI3; Tpn1; troponin 1, type 3; troponin I type 1 (skeletal, slow); troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, skeletal, slow; troponin I, skeletal, slow 1; Troponin I, slow isoform; troponin I, slow skeletal; troponin I, slow skeletal muscle; Troponin I, slow-twitch isoform; troponin I1, slow skeletal type; troponin I3, cardiac type | |
TNNI1, TNNI3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title