Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin I Type 3 (cardiac) Antibody (625), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073085AF594
Description
Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoassay, ELISA, Affinity PurificationSpecifications
Troponin I Type 3 (cardiac) | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoassay, Sandwich ELISA, Immunoaffinity Chromatography | |
625 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoassay, Sandwich ELISA, Immunoaffinity Purification | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human cTn complex. | |
0.2 mL | |
Apoptosis, Cell Biology, Cellular Markers, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
7137 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction