Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin I Type 3 (cardiac) Antibody (MF4cc), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010239FR
Description
Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human, Rat, Porcine, Bovine, Canine, Feline, Goat, Fish (Negative), Rabbit (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Troponin I Type 3 (cardiac) | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNNI3 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with free human cTnI. | |
0.2 mL | |
Apoptosis, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers | |
7137 | |
Human, Rat, Pig, Bovine, Canine, Feline, Goat, Fish (Negative), Rabbit (Negative) | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MF4cc | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
No cross-reaction with skeletal TnT | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction