Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin T type 1 (slow skeletal) Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226095
Description
Troponin T type 1 (slow skeletal) Polyclonal specifically detects Troponin T type 1 (slow skeletal) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA, Single Cell Western.Specifications
Troponin T | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
TNNT1 | |
Peptide with sequence C-EKAQELSDWIHQLE corresponding to internal region according to NP_003274.3, NP_001119604.1, NP_001119605.1. | |
0.1 mg | |
Cytoskeleton Markers, Stem Cell Markers | |
7138 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Western Blot | |
0.5 mg/mL | |
Western Blot 0.03 - 0.1 ug/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:128000, Single Cell Western 100 ug/mL | |
ANM, MGC104241 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize all reported isoforms (NP_003274.3; NP_001119604.1; NP_001119605.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction