Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin T type 1 (slow skeletal) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238855
Description
Troponin T type 1 (slow skeletal) Polyclonal specifically detects Troponin T type 1 (slow skeletal) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Troponin T type 1 (slow skeletal) | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
P13805 | |
TNNT1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EEEEAPEEPEPVAEPEEERPKPSRPVVPPLIPPKIPEGE | |
0.1 mL | |
Cytoskeleton Markers, Stem Cell Markers | |
7138 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ANMtroponin T, slow skeletal muscle, FLJ98147, MGC104241, Slow skeletal muscle troponin T, STNT, TNTS, TNTtroponin T1, skeletal, slow, troponin T type 1 (skeletal, slow), troponin-T1, skeletal, slow | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction