Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin T Type 3 (fast skeletal) Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297579R
Description
Troponin T Type 3 (fast skeletal) Polyclonal specifically detects Troponin T Type 3 (fast skeletal) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Troponin T Type 3 (fast skeletal) | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
Beta-TnTF, fast skeletal muscle, TnTf, troponin T type 3 (skeletal, fast), troponin T3, skeletal, fast, troponin-T3, skeletal, fast | |
Produced in rabbits immunized with E. coli-derived Human Troponin T Type 3 (fast skeletal) fragment. | |
0.1 mL | |
Apoptosis, Cell Biology, Cellular Markers | |
7140 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction