Learn More
Invitrogen™ TROVE2 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HepG2, Molt-4, Raji, mouse heart. Predicted reactivity: Mouse (89%), Rat (88%), Xenopus laevis (80%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TROVE2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RO60 |
| Gene Accession No. | O08848, P10155 |
| Gene Alias | 1810007I17Rik; 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; 60 kDa SS-A/Ro ribonucleoprotein-like protein; A530054J02Rik; AI646302; gastric cancer multi-drug resistance protein; LOW QUALITY PROTEIN: 60 kDa SS-A/Ro ribonucleoprotein; Ro 60 kDa autoantigen; RO60; Ro60 autoantigen; Ro60, Y RNA binding protein; roRNP; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); Ssa; SS-A; SS-A/Ro; SSA2; TROVE domain family member 2; TROVE domain family, member 2; Trove2 |
| Gene Symbols | RO60 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.