Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TROVE2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA564899

Catalog No. PIPA564899


Only null left
Add to Cart

Description

Description

Immunogen sequence: FLLAVDVSASM NQRVLGSILN ASTVAAAMCM VVTRTEKDSY VVAFSDEMVP CPVTTDMTLQ QVLMAMSQIP AGGTDCSLPM IWAQKTNTPA DVFIVFTDNE TFAGGVHPAI ALREYRKKMD IPAKLIVCGM TSNGF Highest antigen sequence identity to the following orthologs - mouse 87%, rat 86%.

TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TROVE2
Polyclonal
Unconjugated
RO60
1810007I17Rik; 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; 60 kDa SS-A/Ro ribonucleoprotein-like protein; A530054J02Rik; AI646302; gastric cancer multi-drug resistance protein; LOW QUALITY PROTEIN: 60 kDa SS-A/Ro ribonucleoprotein; Ro 60 kDa autoantigen; RO60; Ro60 autoantigen; Ro60, Y RNA binding protein; roRNP; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); Ssa; SS-A; SS-A/Ro; SSA2; TROVE domain family member 2; TROVE domain family, member 2; Trove2
Rabbit
Antigen Affinity Chromatography
RUO
6738
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P10155
RO60
Recombinant Human TROVE2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.