Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TROVE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110661 Supplier Invitrogen™ Supplier No. PA5110661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLLAVDVSAS MNQRVLGSIL NASTVAAAMC MVVTRTEKDS YVVAFSDEMV PCPVTTDMTL QQVLMAMSQI PAGGTDCSLP MIWAQKTNTP ADVFIVFTDN ETFAGGVHPA IALREYRKKM DIPAKLIVCG MTSNGF Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 87%.

TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TROVE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RO60
Gene Accession No. P10155
Gene Alias 1810007I17Rik; 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; 60 kDa SS-A/Ro ribonucleoprotein-like protein; A530054J02Rik; AI646302; gastric cancer multi-drug resistance protein; LOW QUALITY PROTEIN: 60 kDa SS-A/Ro ribonucleoprotein; Ro 60 kDa autoantigen; RO60; Ro60 autoantigen; Ro60, Y RNA binding protein; roRNP; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); Ssa; SS-A; SS-A/Ro; SSA2; TROVE domain family member 2; TROVE domain family, member 2; Trove2
Gene Symbols RO60
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TROVE2. Recombinant protein control fragment (Product #RP-101875).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.