Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROVE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 127181AP150UL

Description
TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.Specifications
TROVE2 | |
Polyclonal | |
Unconjugated | |
TROVE2 | |
60 kDa ribonucleoprotein Ro, 60 kDa Ro protein, Ro 60 kDa autoantigen, RO60, RoRNP, Sjoegren syndrome antigen A2, SS A, SSA2, TROVE domain family member 2, TROVE domain family, member 2, TROVE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20822, 304833, 6738 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08848, P10155 | |
TROVE2 | |
TROVE2 Fusion Protein Ag3412 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction