Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TROY Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA529833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529833 Supplier Invitrogen™ Supplier No. MA529833
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TNFRSF19/TROY.

Tumor necrosis factor receptor superfamily member 19, commonly known as TROY, is a gene located on chromosome 1p36.22 that encodes a member of the tumor necrosis factor receptor (TNFR) superfamily. TROY is expressed in the nervous system, skin, and various tissues, where it plays a role in regulating cell apoptosis, inflammation, and development. As a receptor, TROY interacts with ligands and other signaling molecules to modulate pathways involved in cell survival, differentiation, and immune responses. In the context of the nervous system, TROY has been implicated in axonal outgrowth and regeneration, particularly influencing signaling pathways related to neural development and repair. Additionally, TROY has been studied in cancer research due to its potential role in tumor growth and progression, making it a candidate for therapeutic targeting. Its involvement in diverse biological processes highlights its significance in maintaining cellular homeostasis and its potential as a therapeutic target in neurobiology and oncology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TROY
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Tnfrsf19
Gene Accession No. Q9NS68
Gene Alias AL023044; AW123854; RGD1564996; TAJ; TAJ-alpha; TNF receptor superfamily member 19; Tnfrsf19; Toxicity and JNK inducer; TRADE; TROY; tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333
Gene Symbols Tnfrsf19
Host Species Mouse
Immunogen Recombinant Human TNFRSF19/TROY protein (Met1-Leu170).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.