Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROY/TAJ Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501993827
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3827 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-3827 Supplier Active Motif Supplier No. 40937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.

Specifications

Antigen TROY/TAJ
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.5
Gene Tnfrsf19
Gene Accession No. Q9JLL3, Q9NS68
Gene Alias AL023044; AW123854; RGD1564996; Taj; TAJ-alpha; TNF receptor superfamily member 19; TNFRSF19; Toxicity and JNK inducer; TRADE; TROY; Tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333
Gene Symbols Tnfrsf19
Host Species Rabbit
Immunogen This TROY/TAJ antibody was raised against a mixture of synthetic peptides corresponding to amino acid residues 29-44, 204-219, and 409-421 of human TROY/TAJ.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29820, 55504
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.