Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROY/TNFRSF19 Antibody (104), DyLight 650 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290450C
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for ELISA, ELISASpecifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| DyLight 650 | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TROY/TNFRSF19 (NP_001157627.1; Met1-Leu170). | |
| 0.1 mL | |
| Apoptosis | |
| 55504 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 104 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction